4-(2-Thienyl)but-3-en-2-one ≥97%

Supplier: Matrix Scientific

114227-10G 114227-1G 114227-50G
102893-608EA 1820 USD
102893-608 102893-606 102893-604
4-(2-Thienyl)but-3-en-2-one ≥97%
4-(2-Thienyl)but-3-en-2-one

Hazard: Irritant

Formula: C₈H₈OS
MDL Number: MFCD00053051
CAS Number: 874-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR