2,2'-Thenoin ≥97%
Supplier: Matrix Scientific
114226-1G
114226-10G
102893-602EA
1333.85
USD
102893-602
102893-600
2,2'-Thenoin ≥97%
2,2'-Thenoin
Hazard: Irritant
Formula:
C₁₀H₈O₂S₂ |
MDL Number:
MFCD00022505 CAS Number: 27761-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...