N,2-Dimethyl-3-pyridinecarboxamide ≥97%

Supplier: Matrix Scientific

114214-1G
102893-576EA 690.77 USD
102893-576
N,2-Dimethyl-3-pyridinecarboxamide ≥97%
N,2-Dimethyl-3-pyridinecarboxamide

Hazard: Irritant

Formula: C₈H₁₀N₂O
MDL Number: MFCD18384631
CAS Number: 77594-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR