4-(1-Methylethyl)-2-pyridinecarbonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Isopropylpyridine-2-carbonitrile

114206-1G
102893-558EA 2061.54 USD
102893-558
4-(1-Methylethyl)-2-pyridinecarbonitrile ≥97%
4-(1-Methylethyl)-2-pyridinecarbonitrile

Hazard: Irritant

Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD18384514
CAS Number: 676136-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR