3,4-Dimethoxy-2'-hydroxychalcone ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(3,4-Dimethoxyphenyl)-1-(2-hydroxyphenyl)prop-2-en-1-one

114200-1G
102893-546EA 253.85 USD
102893-546
3,4-Dimethoxy-2'-hydroxychalcone ≥97%
3,4-Dimethoxy-2'-hydroxychalcone

Hazard: Irritant

Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
MDL Number: MFCD00017630
CAS Number: 19152-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR