2-Chloro-3-iodophenol ≥97%

Supplier: Matrix Scientific

114194-1G
102893-534EA 1176.92 USD
102893-534
2-Chloro-3-iodophenol ≥97%
2-Chloro-3-iodophenol

Hazard: Irritant

Formula: C₆H₄ClIO
MDL Number: MFCD08166415
CAS Number: 666727-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR