1-[(4-Bromophenyl)sulfanyl]-4-nitrobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Bromophenyl)(4-nitrophenyl)sulphane(4-Bromophenyl)(4-nitrophenyl)sulfane

114166-1G
102893-486EA 690.77 USD
102893-486
1-[(4-Bromophenyl)sulfanyl]-4-nitrobenzene ≥97%
1-[(4-Bromophenyl)sulfanyl]-4-nitrobenzene

Hazard: Irritant

Formula: C₁₂H₈BrNO₂S
MW: 310.18 g/mol
MDL Number: MFCD00172327
CAS Number: 21969-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR