1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 2,6-Difluoro-3-nitrobenzotrifluoride

114154-1G
102893-456EA 300 USD
102893-456
1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene ≥97%
1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₇H₂F₅NO₂
MW: 227.09 g/mol
MDL Number: MFCD12924861
CAS Number: 123973-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR