4-(4-Iodophenoxy)benzonitrile ≥97%

Supplier: Matrix Scientific

114152-1G
102893-452EA 641.54 USD
102893-452
4-(4-Iodophenoxy)benzonitrile ≥97%
4-(4-Iodophenoxy)benzonitrile

Hazard: Irritant

Formula: C₁₃H₈INO
MDL Number: MFCD11102672
CAS Number: 856704-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR