3-(4-Chlorophenoxy)pentane-2,4-dione ≥97%

Supplier: Matrix Scientific

114147-1G
102893-442EA 232.31 USD
102893-442
3-(4-Chlorophenoxy)pentane-2,4-dione ≥97%
3-(4-Chlorophenoxy)pentane-2,4-dione

Hazard: Irritant

Formula: C₁₁H₁₁ClO₃
MDL Number: MFCD00204239
CAS Number: 31168-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR