Allyl-4-oxopiperidine-1-carboxylate ≥97%

Synonyms: 1-N-Alloc-4-piperidone

114136-1G 114136-10G
102893-416EA 206.15 USD
102893-416 102893-418
Allyl-4-oxopiperidine-1-carboxylate ≥97%
Allyl-4-oxopiperidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
MDL Number: MFCD02183578
CAS Number: 306296-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR