tert-Butyl-2-ethylpiperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

114134-1G
102893-412EA 641.54 USD
102893-412
tert-Butyl-2-ethylpiperidine-1-carboxylate ≥97%
tert-Butyl-2-ethylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₃NO₂
MW: 213.32 g/mol
MDL Number: MFCD14636609
CAS Number: 409061-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR