2-Chloro-N-piperidin-4-ylpyrimidin-4-amine ≥97%
Supplier: Matrix Scientific
114132-1G
102893-406EA
813.85
USD
102893-406
2-Chloro-N-piperidin-4-ylpyrimidin-4-amine ≥97%
2-Chloro-N-piperidin-4-ylpyrimidin-4-amine
Hazard: Irritant
Formula:
C₉H₁₃ClN₄ |
MDL Number:
MFCD09756895 CAS Number: 945895-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...