4-Methyl-1,2,3-thiadiazole-5-carboxamide ≥97%

Supplier: Matrix Scientific

114126-1G
102893-364EA 183.08 USD
102893-364
4-Methyl-1,2,3-thiadiazole-5-carboxamide ≥97%
4-Methyl-1,2,3-thiadiazole-5-carboxamide

Hazard: Irritant

Formula: C₄H₅N₃OS
MDL Number: MFCD00052214
CAS Number: 175136-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR