2-(1-Methylhydrazino)quinoxaline ≥97%

Supplier: Matrix Scientific

114124-1G
102893-360EA 303.08 USD
102893-360
2-(1-Methylhydrazino)quinoxaline ≥97%
2-(1-Methylhydrazino)quinoxaline

Hazard: Irritant

Formula: C₉H₁₀N₄
MDL Number: MFCD00085138
CAS Number: 16621-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR