4-Hydroxy-7-nitroquinoline ≥97%

Supplier: Matrix Scientific

114122-1G
102893-356EA 813.85 USD
102893-356
4-Hydroxy-7-nitroquinoline ≥97%
4-Hydroxy-7-nitroquinoline

Hazard: Irritant

Formula: C₉H₆N₂O₃
MDL Number: MFCD08688614
CAS Number: 6270-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR