1-Methyl-2-phenylbenzimidazole ≥97%

Supplier: Matrix Scientific

114116-1G
102893-342EA 813.85 USD
102893-342
1-Methyl-2-phenylbenzimidazole ≥97%
1-Methyl-2-phenylbenzimidazole

Hazard: Irritant

Formula: C₁₄H₁₂N₂
MDL Number: MFCD00005764
CAS Number: 2622-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR