1-(Chloromethyl)-4-propoxybenzene ≥97%

Supplier: Matrix Scientific

114109-1G
102893-334EA 813.85 USD
102893-334
1-(Chloromethyl)-4-propoxybenzene ≥97%
1-(Chloromethyl)-4-propoxybenzene

Hazard: Irritant

Formula: C₁₀H₁₃ClO
MDL Number: MFCD11121524
CAS Number: 40141-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR