Cyclohexanemethyl isocyanate ≥97%

Supplier: Matrix Scientific

114106-1G
102893-328EA 447.69 USD
102893-328
Cyclohexanemethyl isocyanate ≥97%
Cyclohexanemethyl isocyanate

Hazard: Toxic/Stored under Argon/Moisture Sensitive/Refrigerate

Formula: C₈H₁₃NO
Boiling Pt: 193…194 °C
Density: 0.974
Flash Pt: 76 °C
MDL Number: MFCD03701631
CAS Number: 51163-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR