(4-Methyl-1,2,3-thiadiazol-5-yl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 5-(Hydroxymethyl)-4-methyl-1,2,3-thiadiazole

114098-1G
102893-306EA 193.85 USD
102893-306
(4-Methyl-1,2,3-thiadiazol-5-yl)methanol ≥97%
(4-Methyl-1,2,3-thiadiazol-5-yl)methanol

Hazard: Irritant

Formula: C₄H₆N₂OS
MW: 130.17 g/mol
MDL Number: MFCD07371384
CAS Number: 163008-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR