2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan ≥97%

Supplier: Matrix Scientific

114097-1G
102893-304EA 813.85 USD
102893-304
2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan ≥97%
2,3-Dihydro-5-(2-hydroxyethyl)benzo[b]furan

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD06797643
CAS Number: 87776-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR