4-(1-Hydroxyethyl)pyridine ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-(Pyridin-4-yl)ethanol
, 1-(Pyridin-4-yl)ethan-1-ol
114094-1G
102893-298EA
329.23
USD
102893-298
4-(1-Hydroxyethyl)pyridine ≥97%
4-(1-Hydroxyethyl)pyridine
Formula:
C₇H₉NO MW: 123.15 g/mol Melting Pt: 51…55 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077867 CAS Number: 23389-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...