(4-Propoxyphenyl)methanol ≥97%

Supplier: Matrix Scientific

114089-1G
102893-288EA 620 USD
102893-288
(4-Propoxyphenyl)methanol ≥97%
(4-Propoxyphenyl)methanol

Hazard: Irritant

Formula: C₁₀H₁₄O₂
MDL Number: MFCD06202634
CAS Number: 90925-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR