3-Bromo-2,2-dimethyl-1-propanol ≥97%

Supplier: Matrix Scientific

114084-10G
102893-280EA 376.92 USD
102893-280
3-Bromo-2,2-dimethyl-1-propanol ≥97%
3-Bromo-2,2-dimethyl-1-propanol

Hazard: Irritant/Light Sensitive

Formula: C₅H₁₁BrO
Boiling Pt: 184…187 °C
Density: 1.358
Flash Pt: 76 °C
MDL Number: MFCD00004683
CAS Number: 40894-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR