6-Bromo-3-methyl-3,4-dihydro-2(1H)-quinazolinone ≥97%

Supplier: Matrix Scientific

114080-1G
102893-270EA 813.85 USD
102893-270
6-Bromo-3-methyl-3,4-dihydro-2(1H)-quinazolinone ≥97%
6-Bromo-3-methyl-3,4-dihydro-2(1H)-quinazolinone

Hazard: Irritant

Formula: C₉H₉BrN₂O
MDL Number: MFCD02625685
CAS Number: 328956-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR