3,4-Dichloropyridazine ≥97%
Supplier: Matrix Scientific
114076-1G
102893-260EA
1927.69
USD
102893-260
3,4-Dichloropyridazine ≥97%
3,4-Dichloropyridazine
Hazard: Irritant
Formula:
C₄H₂Cl₂N₂ |
MDL Number:
MFCD20040517 CAS Number: 1677-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...