4-Chloro-2-[(pyrrolidin-1-yl)carbonyl]pyridine ≥97%
Supplier: Matrix Scientific
114069-1G
102893-246EA
813.85
USD
102893-246
4-Chloro-2-[(pyrrolidin-1-yl)carbonyl]pyridine ≥97%
4-Chloro-2-[(pyrrolidin-1-yl)carbonyl]pyridine
Hazard: Irritant
Formula:
C₁₀H₁₁ClN₂O |
MDL Number:
MFCD11177040 CAS Number: 260783-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...