7-Fluoro-1,2,3,4-tetrahydroisoquinoline ≥97%

Supplier: Matrix Scientific

114060-1G
102898-362EA 958.46 USD
102898-362
7-Fluoro-1,2,3,4-tetrahydroisoquinoline ≥97%
7-Fluoro-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₉H₁₀FN
MW: 151.181 g/mol
MDL Number: MFCD03094746
CAS Number: 406923-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR