Ethyl-4-[cyclopropyl(hydroxy)methylene]-3,5-dioxocyclohexanecarboxylate ≥97%

Supplier: Matrix Scientific

114027-1G
102893-166EA 690.77 USD
102893-166
Ethyl-4-[cyclopropyl(hydroxy)methylene]-3,5-dioxocyclohexanecarboxylate ≥97%
Ethyl-4-[cyclopropyl(hydroxy)methylene]-3,5-dioxocyclohexanecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆O₅
MDL Number: MFCD01632318
CAS Number: 95266-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR