Ethyl cyclopentylideneacetate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-cyclopentylideneacetate

114026-1G
102893-164EA 690.77 USD
102893-164
Ethyl cyclopentylideneacetate ≥97%
Ethyl cyclopentylideneacetate

Hazard: Irritant

Formula: C₉H₁₄O₂
MW: 154.21 g/mol
MDL Number: MFCD11976268
CAS Number: 1903-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR