Methyl cyclobutylideneacetate ≥97%

Supplier: Matrix Scientific

114019-1G
102893-150EA 544.62 USD
102893-150
Methyl cyclobutylideneacetate ≥97%
Methyl cyclobutylideneacetate

Hazard: Flammable/Toxic

Formula: C₇H₁₀O₂
MDL Number: MFCD12405321
CAS Number: 143140-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR