Methyl-1-benzyl-3-pyrrolidineacetate ≥97%

Supplier: Matrix Scientific

114014-1G
102893-140EA 884.62 USD
102893-140
Methyl-1-benzyl-3-pyrrolidineacetate ≥97%
Methyl-1-benzyl-3-pyrrolidineacetate

Hazard: Irritant

Formula: C₁₄H₁₉NO₂
MDL Number: MFCD04038689
CAS Number: 95274-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR