Methyl (4-benzylpiperazin-1-yl)propionate ≥97%

Supplier: Matrix Scientific

114009-1G
102893-130EA 813.85 USD
102893-130
Methyl (4-benzylpiperazin-1-yl)propionate ≥97%
Methyl (4-benzylpiperazin-1-yl)propionate

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD02684217
CAS Number: 204059-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR