Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 1,2,3,4-tetrahydroquinoline-6-carboxylate

114006-1G
102893-122EA 473.85 USD
102893-122
Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate ≥97%
Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MW: 191.23 g/mol
MDL Number: MFCD08692534
CAS Number: 177478-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR