Methyl-3-iodo-1H-indole-6-carboxylate ≥97%

Supplier: Matrix Scientific

114004-1G
102893-118EA 350.77 USD
102893-118
Methyl-3-iodo-1H-indole-6-carboxylate ≥97%
Methyl-3-iodo-1H-indole-6-carboxylate

Hazard: Irritant

Formula: C₁₀H₈INO₂
MW: 301.09 g/mol
MDL Number: MFCD06797504
CAS Number: 850374-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR