Methyl-2-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetate ≥97%

Supplier: Matrix Scientific

113998-1G
102893-104EA 958.46 USD
102893-104
Methyl-2-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetate ≥97%
Methyl-2-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetate

Hazard: Irritant

Formula: C₈H₁₀N₂O₄
MDL Number: MFCD00457632
CAS Number: 134456-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR