Methyl-4-nitro-1H-pyrazole-3-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 4-nitro-1H-pyrazole-5-carboxylateMethyl 4-nitro-1H-pyrazole-3-carboxylate

113995-1G
102893-098EA 23.08 USD
102893-098
Methyl-4-nitro-1H-pyrazole-3-carboxylate ≥97%
Methyl-4-nitro-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₅H₅N₃O₄
MW: 171.11 g/mol
MDL Number: MFCD00505604
CAS Number: 138786-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR