Ethyl-2-phenyl-1,3-thiazole-4-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-phenylthiazole-4-carboxylate
113991-1G
102898-360EA
400
USD
102898-360
Ethyl-2-phenyl-1,3-thiazole-4-carboxylate ≥97%
Ethyl-2-phenyl-1,3-thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₁NO₂S MW: 233.29 g/mol |
MDL Number:
MFCD06205115 CAS Number: 59937-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...