Methyl-3-amino-2-methylpropanoate ≥95%

Supplier: Matrix Scientific

113988-1G
102898-354EA 958.46 USD
102898-354
Methyl-3-amino-2-methylpropanoate ≥95%
Methyl-3-amino-2-methylpropanoate

Hazard: Irritant

Formula: C₅H₁₁NO₂
MDL Number: MFCD12132810
CAS Number: 14678-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR