Ethyl-4-(benzyloxy)-3-oxobutanoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-(benzyloxy)-3-oxobutanoate

113987-1G
102898-352EA 864.62 USD
102898-352
Ethyl-4-(benzyloxy)-3-oxobutanoate ≥97%
Ethyl-4-(benzyloxy)-3-oxobutanoate

Hazard: Irritant

Formula: C₁₃H₁₆O₄
MW: 236.27 g/mol
MDL Number: MFCD19388798
CAS Number: 67354-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR