Methyl-5-formyl-2-furoate ≥95%

Supplier: Matrix Scientific

113976-1G
102893-066EA 735.38 USD
102893-066
Methyl-5-formyl-2-furoate ≥95%
Methyl-5-formyl-2-furoate

Hazard: Irritant

Formula: C₇H₆O₄
MDL Number: MFCD06803263
CAS Number: 5904-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR