Ethyl-3-amino-4-(isopropylsulfonyl)-5-(methylthio)thiophene-2-carboxylate ≥97%

Supplier: Matrix Scientific

113973-1G
102898-342EA 183.08 USD
102898-342
Ethyl-3-amino-4-(isopropylsulfonyl)-5-(methylthio)thiophene-2-carboxylate ≥97%
Ethyl-3-amino-4-(isopropylsulfonyl)-5-(methylthio)thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₇NO₄S₃
MDL Number: MFCD00085052
CAS Number: 175202-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR