Methyl pyridin-3-ylacetate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(pyridin-3-yl)acetate

113966-1G
102898-104EA 570.77 USD
102898-104
Methyl pyridin-3-ylacetate ≥97%
Methyl pyridin-3-ylacetate

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00023617
CAS Number: 39998-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR