Ethyl-2-benzoylpropionate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-methyl-3-oxo-3-phenylpropanoate2-Benzoylpropionic Acid Ethyl Ester

113957-1G
102898-086EA 958.46 USD
102898-086
Ethyl-2-benzoylpropionate ≥97%
Ethyl-2-benzoylpropionate

Purity: 97%
CAS: 10488-87-6
MDL Number: MFCD00026882
Molecular Formula: C12H14O3
Molecular Weight: 206,24
Hazard: Irritant

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD00026882
CAS Number: 10488-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR