Methyl-3-(2,4-difluorophenyl)-3-oxopropanoate ≥97%

Supplier: Matrix Scientific

113956-1G
102893-058EA 813.85 USD
102893-058
Methyl-3-(2,4-difluorophenyl)-3-oxopropanoate ≥97%
Methyl-3-(2,4-difluorophenyl)-3-oxopropanoate

Hazard: Irritant

Formula: C₁₀H₈F₂O₃
MDL Number: MFCD03424792
CAS Number: 185302-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR