Methyl-2-(3,4-dimethoxyphenyl)acetate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-(3,4-dimethoxyphenyl)acetate
113954-1G
113954-10G
102893-052EA
1090.77
USD
102893-052
102893-050
Methyl-2-(3,4-dimethoxyphenyl)acetate ≥97%
Methyl-2-(3,4-dimethoxyphenyl)acetate
Hazard: Irritant
Formula:
C₁₁H₁₄O₄ MW: 210.23 g/mol |
MDL Number:
MFCD00017206 CAS Number: 15964-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...