Methyl-3-iodo-5-nitrobenzoate ≥97%

Supplier: Matrix Scientific

113951-1G
102898-082EA 738.46 USD
102898-082
Methyl-3-iodo-5-nitrobenzoate ≥97%
Methyl-3-iodo-5-nitrobenzoate

Hazard: Irritant

Formula: C₈H₆INO₄
MDL Number: MFCD00463865
CAS Number: 50765-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR