Methyl-2-bromo-3-(trifluoromethyl)benzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-bromo-3-(trifluoromethyl)benzoate

113924-1G
102893-006EA 492.31 USD
102893-006
Methyl-2-bromo-3-(trifluoromethyl)benzoate ≥97%
Methyl-2-bromo-3-(trifluoromethyl)benzoate

Hazard: Irritant

Formula: C₉H₆BrF₃O₂
MW: 283.05 g/mol
MDL Number: MFCD14697994
CAS Number: 1214362-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR