Methyl-3-(2-methyl-thiazol-4-yl)benzoate ≥97%

Supplier: Matrix Scientific

113922-1G
102893-002EA 426.15 USD
102893-002
Methyl-3-(2-methyl-thiazol-4-yl)benzoate ≥97%
Methyl-3-(2-methyl-thiazol-4-yl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₁NO₂S
MDL Number: MFCD06797464
CAS Number: 850375-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR