Methyl-2-chloro-4,5-difluorobenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-4,5-difluorobenzoate2-Chloro-4,5-difluorobenzoic acid, methyl ester

113920-1G
102892-998EA 738.46 USD
102892-998
Methyl-2-chloro-4,5-difluorobenzoate ≥97%
Methyl-2-chloro-4,5-difluorobenzoate

Hazard: Irritant

Formula: C₈H₅ClF₂O₂
MW: 206.58 g/mol
MDL Number: MFCD10566827
CAS Number: 128800-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR